4'-Chloropropiophenone

Supplier: TCI America

Synonyms: p-Chloropropiophenone4-Chloropropiophenone

C1616-25G C1616-500G
TCC1616-500GEA 249.75 USD
TCC1616-500G TCC1616-025G
4'-Chloropropiophenone
4'-Chloropropiophenone

CAS Number: 6285-05-8
MDL Number: MFCD00000626
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Form: Crystal
Color: White
Boiling point (°C): 152
Melting point (°C): 35
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Chlorophenyl Ethyl Ketone

Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 152 °C
Melting Pt: 35 °C
Flash Pt: 110 °C
MDL Number: MFCD00000626
CAS Number: 6285-05-8
EINECS: 228-511-6
https://us.vwr.com/stibo/web/std.lang.all/88/34/28938834.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR