1-Chloro-2-octyne ≥97.0%
Supplier: TCI America
C1480-25ML
TCC1480-025MLEA
137.92
USD
TCC1480-025ML
1-Chloro-2-octyne ≥97.0%
1-Chloro-2-octyne
CAS Number: 51575-83-8
MDL Number: MFCD00274223
Molecular Formula: C8H13Cl
Molecular Weight: 144.64
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 68
Specific Gravity (20/20): 0.93
Formula:
C₈H₁₃Cl MW: 144.64 g/mol Boiling Pt: 81 °C Density: 0.93 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00274223 CAS Number: 51575-83-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...