TOKYC1470

C1470-1G
TCC1470-001GEA 61.39 USD
TCC1470-001G
TOKYC1470
CAS Number: 293-96-9
MDL Number: MFCD00003711
Molecular Formula: C10H20
Molecular Weight: 140.27
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Melting point (°C): 10
Flash Point (°C): 65
Specific Gravity (20/20): 0.86
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR