4-Nitrobenzyl chloroformate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Nitrobenzyl carbonochloridate

C1077-25G
TCC1077-025GEA 300 USD
TCC1077-025G
4-Nitrobenzyl chloroformate ≥97.0% (by titrimetric analysis)
4-Nitrobenzyl chloroformate

CAS Number: 4457-32-3
MDL Number: MFCD00007375
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 34
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chloroformic Acid 4-Nitrobenzyl Ester

Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
Boiling Pt: 230 °C
Melting Pt: 34 °C
Flash Pt: 110 °C
MDL Number: MFCD00007375
CAS Number: 4457-32-3
EINECS: 224-708-6
https://us.vwr.com/stibo/web/std.lang.all/83/40/28938340.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR