Methyl-4-chloroacetoacetate ≥96.0%
Supplier: TCI America
Synonyms:
Methyl 4-chloro-3-oxobutanoate
C0910-25ML
C0910-250ML
TCC0910-025MLEA
45.54
USD
TCC0910-025ML
TCC0910-250ML
Methyl-4-chloroacetoacetate ≥96.0%
Methyl 4-chloroacetoacetate
CAS Number: 32807-28-6
MDL Number: MFCD00000938
Molecular Formula: C5H7ClO3
Molecular Weight: 150.56
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 97
Melting point (°C): 14
Flash Point (°C): 108
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: 4-Chloroacetoacetic Acid Methyl Ester
Formula:
C₅H₇ClO₃ MW: 150.56 g/mol Boiling Pt: 97 °C Melting Pt: 14 °C Density: 1.28 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00000938 CAS Number: 32807-28-6 EINECS: 251-230-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...