2-Chloroheptane ≥97.0%

Supplier: TCI America

Synonyms: 2-Heptyl chloride

C0852-5ML C0852-25ML
TCC0852-025MLEA 381.53 USD
TCC0852-025ML TCC0852-005ML
2-Chloroheptane ≥97.0%
2-Chloroheptane

CAS Number: 1001-89-4
MDL Number: MFCD00039353
Molecular Formula: C7H15Cl
Molecular Weight: 134.65
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 2-Heptyl Chloride

Formula: C₇H₁₅Cl
MW: 134.65 g/mol
Boiling Pt: 146 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00039353
CAS Number: 1001-89-4
EINECS: 213-683-7
https://us.vwr.com/stibo/web/std.lang.all/79/95/28937995.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR