Cresol (mixture of isomers) ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Hydroxy(methyl)benzene
, Methylphenol
, Hydroxytoluene
C0698-500ML
C0698-25ML
TCC0698-500MLEA
158.54
USD
TCC0698-500ML
TCC0698-025ML
Cresol (mixture of isomers) ≥99.0%
Cresol (mixture of isomers)
CAS Number: 1319-77-3
MDL Number: MFCD00151099
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 192
Flash Point (°C): 81
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: Cresylic Acid, Tricresol, Methylphenol
Formula:
C₇H₈O MW: 108.14 g/mol Boiling Pt: 192 °C Density: 1.04 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00151099 CAS Number: 1319-77-3 EINECS: 215-293-2 Merck Index: 12,02645 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...