Cycloheptanol ≥98.0%

Supplier: TCI America

Synonyms: HydroxycycloheptaneSuberyl alcohol

C0678-25ML
TCC0678-025MLEA 59 USD
TCC0678-025ML
Cycloheptanol ≥98.0%
Cycloheptanol

CAS Number: 502-41-0
MDL Number: MFCD00004150
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 71
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Hydroxycycloheptane, Suberyl Alcohol

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 185 °C
Melting Pt: 2 °C
Density: 0.96 (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004150
CAS Number: 502-41-0
EINECS: 207-936-0
https://us.vwr.com/stibo/web/std.lang.all/77/17/28937717.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR