2-Chlorocyclohexanol ≥95.0%

Supplier: TCI America

Synonyms: Cyclohexene chlorohydrin

C0656-5G C0656-25G
TCC0656-025GEA 332.84 USD
TCC0656-025G TCC0656-005G
2-Chlorocyclohexanol ≥95.0%
2-Chlorocyclohexanol

CAS Number: 1561-86-0
MDL Number: MFCD00003860
Molecular Formula: C6H11ClO
Molecular Weight: 134.60
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Boiling point (°C): 84
Flash Point (°C): 70
Freezing point (°C): 23


Other Product Names / Synonyms: Cyclohexene Chlorohydrin

Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 84 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00003860
CAS Number: 1561-86-0
EINECS: 216-339-4
https://us.vwr.com/stibo/web/std.lang.all/76/95/28937695.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR