4-Chlorobutyric acid ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Chlorobutanoic acid
C0651-25G
C0651-500G
TCC0651-025GEA
64.94
USD
TCC0651-025G
TCC0651-500G
4-Chlorobutyric acid ≥95.0% (by titrimetric analysis)
4-Chlorobutyric acid
CAS Number: 627-00-9
MDL Number: MFCD00002818
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 112
Freezing point (°C): 14
Specific Gravity (20/20): 1.23
Formula:
C₄H₇ClO₂ MW: 122.55 g/mol Boiling Pt: 125 °C Melting Pt: 16 °C Density: 1.23 (20 °C) Flash Pt: 112 °C |
MDL Number:
MFCD00002818 CAS Number: 627-00-9 EINECS: 210-977-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...