1-Chlorodecane ≥97.0%

Supplier: TCI America

C0600-500ML C0600-25ML
TCC0600-025MLEA 46.69 USD
TCC0600-025ML TCC0600-500ML
1-Chlorodecane ≥97.0%
1-Chlorodecane

CAS Number: 1002-69-3
MDL Number: MFCD00000956
Molecular Formula: C10H21Cl
Molecular Weight: 176.73
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Melting point (°C): -34
Flash Point (°C): 102
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Decyl Chloride

Formula: C₁₀H₂₁Cl
MW: 176.73 g/mol
Boiling Pt: 223 °C
Melting Pt: –34 °C
Density: 0.87 (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00000956
CAS Number: 1002-69-3
EINECS: 213-691-0
https://us.vwr.com/stibo/web/std.lang.all/76/40/28937640.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR