(Bromomethyl)trimethylsilane ≥97.0%

Supplier: TCI America

Synonyms: (Trimethylsilyl)methyl bromide

B1892-5G B1892-25G
TCB1892-25GEA 315.67 USD
TCB1892-25G TCB1892-005G
(Bromomethyl)trimethylsilane ≥97.0%
(Bromomethyl)trimethylsilane

CAS Number: 18243-41-9
MDL Number: MFCD00000167
Molecular Formula: C4H11BrSi
Molecular Weight: 167.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Flash Point (°C): 16
Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Trimethylsilyl)methyl Bromide

Formula: BrH₂CSi(CH₃)₃
MW: 167.12 g/mol
Boiling Pt: 117 °C
Density: 1.17 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00000167
CAS Number: 18243-41-9
https://us.vwr.com/stibo/web/std.lang.all/36/62/28933662.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR