5-Bromo-2-methylbenzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 2-Methyl-5-bromobenzothiazole

B1798-5G B1798-25G
TCB1798-005GEA 114.66 USD
TCB1798-005G TCB1798-25G
5-Bromo-2-methylbenzothiazole ≥98.0%
5-Bromo-2-methylbenzothiazole

CAS Number: 63837-11-6
MDL Number: MFCD00603096
Molecular Formula: C8H6BrNS
Molecular Weight: 228.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 79
Storage Temperature: 0-10°C

Formula: C₈H₆BrNS
MW: 228.11 g/mol
Boiling Pt: 170 °C
Melting Pt: 79 °C
MDL Number: MFCD00603096
CAS Number: 63837-11-6
EINECS: 264-510-7
https://us.vwr.com/stibo/web/std.lang.all/35/73/28933573.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR