3-Benzylpyridine ≥98.0%

Supplier: TCI America

B1553-10G
TCB1553-010GEA 285.09 USD
TCB1553-010G
3-Benzylpyridine ≥98.0%
3-Benzylpyridine

CAS Number: 620-95-1
MDL Number: MFCD00006408
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Freezing point (°C): 30

Formula: C₁₂H₁₁N
MW: 169.23 g/mol
MDL Number: MFCD00006408
CAS Number: 620-95-1
EINECS: 210-661-9
https://us.vwr.com/stibo/web/std.lang.all/33/21/28933321.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR