1,4-Bis(1,2-dibromoethyl)benzene ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: alpha,alpha',beta,beta'-Tetrabromo-1,4-diethylbenzene

B1504-5G
TCB1504-005GEA 41.6 USD
TCB1504-005G
1,4-Bis(1,2-dibromoethyl)benzene ≥97.0% (by GC, titration analysis)
1,4-Bis(1,2-dibromoethyl)benzene

CAS Number: 25393-98-0
MDL Number: MFCD00017881
Molecular Formula: C10H10Br4
Molecular Weight: 449.81
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 159


Other Product Names / Synonyms: alpha,alpha',beta,beta'-Tetrabromo-1,4-diethylbenzene

Formula: C₁₀H₁₀Br₄
MW: 449.81 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00017881
CAS Number: 25393-98-0
EINECS: 246-936-5

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR