4-sec-Butylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-(sec-Butyl)aniline
B1256-25ML
TCB1256-025MLEA
316.59
USD
TCB1256-025ML
4-sec-Butylaniline ≥98.0% (by GC, titration analysis)
4-sec-Butylaniline
CAS Number: 30273-11-1
MDL Number: MFCD00010223
Molecular Formula: C10H15N
Molecular Weight: 149.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 238
Flash Point (°C): 107
Specific Gravity (20/20): 0.94
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 238 °C Density: 0.94 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00010223 CAS Number: 30273-11-1 EINECS: 250-108-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...