1,1'-Binaphthyl ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,1'-Binaphthalene
B0805-5G
B0805-1G
TCB0805-001GEA
47.71
USD
TCB0805-001G
TCB0805-5G
1,1'-Binaphthyl ≥98.0%
1,1'-Binaphthyl
CAS Number: 604-53-5
MDL Number: MFCD00041740
Molecular Formula: C20H14
Molecular Weight: 254.33
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: 1,1'-Dinaphthyl
Formula:
C₂₀H₁₄ MW: 254.33 g/mol Boiling Pt: 405 °C (1013 hPa) Melting Pt: 142…144 °C Density: 1.19 g/cm³ (20 °C) |
MDL Number:
MFCD00041740 CAS Number: 604-53-5 EINECS: 210-070-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...