2-Bromo-2-methylbutane ≥95.0%

Supplier: TCI America

B0612-25G
TCB0612-025GEA 143.08 USD
TCB0612-025G
2-Bromo-2-methylbutane ≥95.0%
2-Bromo-2-methylbutane

CAS Number: 507-36-8
MDL Number: MFCD00000127
Molecular Formula: C5H11Br
Molecular Weight: 151.05
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 5
Specific Gravity (20/20): 1.20
Storage Temperature: 0-10°C


Other Product Names / Synonyms: tert-Amyl Bromide

Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 107 °C
Density: 1.20 (20 °C)
Flash Pt: 5 °C
MDL Number: MFCD00000127
CAS Number: 507-36-8
EINECS: 208-071-1
UN: 1993
ADR: 3,II
Merck Index: 12,00644
https://us.vwr.com/stibo/web/std.lang.all/25/46/28932546.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR