Phenylarsonic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B0021-25G
TCB0021-025GEA 196.85 USD
TCB0021-025G
Phenylarsonic acid ≥99.0% (by titrimetric analysis)
Phenylarsonic acid

CAS Number: 98-05-5
MDL Number: MFCD00002097
Molecular Formula: C6H7AsO3
Molecular Weight: 202.04
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: Benzenearsonic Acid

Formula: C₆H₇AsO₃
MW: 202.04 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00002097
CAS Number: 98-05-5
EINECS: 202-631-9
UN: 3465
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/21/96/28932196.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR