Acetoxyacetyl chloride ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Acetoxyacetylchloride2-Chloro-2-oxoethyl acetate

A1500-100G A1500-25G
TCA1500-100GEA 196.85 USD
TCA1500-100G TCA1500-025G
Acetoxyacetyl chloride ≥95.0% (by GC, titration analysis)
Acetoxyacetyl chloride

CAS Number: 13831-31-7
MDL Number: MFCD00011535
Molecular Formula: C4H5ClO3
Molecular Weight: 136.53
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 71
Specific Gravity (20/20): 1.27

Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 160 °C
Density: 1.27 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00011535
CAS Number: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/95/47/28929547.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR