Cyclohexyl acetate ≥99.0%

Supplier: TCI America

Synonyms: Acetic acid cyclohexyl ester

A0028-500ML A0028-25ML
TCA0028-500MLEA 86.28 USD
TCA0028-500ML TCA0028-025ML
Cyclohexyl acetate ≥99.0%
Cyclohexyl acetate

CAS Number: 622-45-7
MDL Number: MFCD00003850
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 58
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Acetic Acid Cyclohexyl Ester

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 174 °C
Melting Pt: –77 °C
Density: 0.97 (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00003850
CAS Number: 622-45-7
EINECS: 210-736-6
UN: 2243
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/74/66/28927466.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR