Potassium peroxodisulfate 5% (w/v) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Dipotassium peroxodisulphate
R6450000-1C
RC6450-32EA
105.04
USD
RC6450-32
Potassium peroxodisulfate 5% (w/v) in aqueous solution
Potassium peroxodisulphate
Formula:
K₂S₂O₈ MW: 270.32 g/mol |
MDL Number:
MFCD00011386 CAS Number: 7727-21-1 UN: 1492 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...