Chlorine water saturated ∼0.3% (w/v)

Warning

R1980000-1C1 R1980000-500C1
RC198016EA 78.71 USD
RC198016 RC198032
Chlorine water saturated ∼0.3% (w/v)
Chlorine water
Melting Pt: 0.0 °C
Density: 1.00 g/cm³
Storage Temperature: Ambient
CAS Number: 7782-50-5
UN: 1760
ADR: 2,

Order Now

Specification Test Results

Appearance Yellow-green liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR