VESTEX™ Active Barrier Protective Lab Coat, Vestagen Technical Textiles
VESTEX™
VT401WHSM
89260-230CS 3236.56 USD
89260-230
VESTEX™ Active Barrier Protective Lab Coat, Vestagen Technical Textiles
Workwear
VESTEX™ Active Barrier protective lab coats are engineered to protect users from unexpected splashes and splatters of blood and bodily fluids.
  • Fabric repels fluids and dirt, and resists exterior stains to keep users clean and dry
  • Breathable 80% polyester/20% cotton construction
  • Side slits are included for convenient pants pocket access
  • Machine washable; can also be autoclaved
  • Unisex Sizing
Lab coat sizes are based on a 96.5cm (38”) center back length on size medium. All lab coats feature 4 button front closure, with traditional styling details such as a notched lapel and a center back belt. The left patch pocket features scissors/utility loops inside, the right patch pocket has a kangaroo pouch, and the left chest pocket has a pen holder.
Comfortable enough to wear all day, VESTEX™ is clinically proven to rapidly prevent, reduce, or eliminate the acquisition and retention of contaminants on the fabric. Fluids literally bead up and roll off of the breathable, comfortable, and durable VESTEX™ fabric. VESTEX™ also contains an antimicrobial element to prevent degradation of the fabric and to control odors caused by bacteria