Flow Cup Viscosity Standards, Fungilab
Supplier: Fungilab
V12C350 V12C200
89237-172EA 206.74 USD
89237-172 89237-170
Flow Cup Viscosity Standards, Fungilab
Standards Viscometry Standards
ISO 17025 / ISO Guide 34 certified flow cup viscosity standards are specifcally formulated for use with viscosity flow cups such as Ford, Shell, Zahn.
Certifications: Certified at 20°C and 25°C, each value is tested to ASTM D 2162 for viscosity measurement and ASTM D 1480 for density measurement. Each value at every test temperature is tested to ASTM D 2162 and are NOT extrapolations. ISO 17025 / ISO 17034 dual certified flow cup standards are especially designed for use in DIN, Ford, ISO, Shell and Zahn flow cups and are also dual purpose density standards.