U-Tube Reverse Flow BS/IP/RF Capillary Viscometers, Fungilab
Supplier: Fungilab
CV006-001 CV006-009 CV006-010 CV006-006 CV006-004 CV006-011 CV006-002 CV006-005 CV006-007 CV006-003 CV006-008
89236-950EA 360.33 USD
89236-950 89236-940 89236-930 89236-942 89236-932 89236-944 89236-934 89236-946 89236-936 89236-948 89236-938
U-Tube Reverse Flow BS/IP/RF Capillary Viscometers, Fungilab
Viscometers Capillary Viscometers
BS/IP/ U Tube Reverse Flow capillary viscosmeter for opaque liquids
Made of borosilicate glass, and non-alterable timing mark.
Certifications: BS, UNE 400313, ASTM D-445, D446, ISO 3104, ISO 3105.
Ordering information: Includes constant value calibration certificate at 40ºC (104ºF) and 100ºC (212ºF).