Digital Rotational Viscometer SMART Series, Fungilab
Supplier: Fungilab
V211002 V210002 V210001 V211001
89237-190EA 6910.88 USD
89237-190 89237-108 89237-110 89237-188
Digital Rotational Viscometer SMART Series, Fungilab
Viscometers Rotational Viscometers
Smart Series is a advanced digital rotational viscometer for viscosity readings
Main features include: 4LCD display for easy menu access and results reading, relative, absoluteand kinematic viscosity reading automatically, unit convert SI to CGS; Autotest and Autorange functions; program features time to torque and time to stop; temperature reading PT100 (optional); Interface: USB connection; 0 working memories; 10 language options. Datalogger software included for downloading data to computer to work in Excel® file (with USB connector). 6 touch keypad. LCD screen. 4 lines.
Indispensable for QC and R&D laboratories.
Certifications: CE certified. TURV certified. ISO 9001. BS: 6075, 5350. ISO: 2555, 1652 . ASTM: 115, 789, 1076, 1084, 1286, 1417, 1439, 1638, 1824, 2196, 2336, 2364, 2393, 2556, 2669, 2849, 2983, 2994, 3232, 3236, 3716.
Ordering information: Standard delivery includes:complete with standard spindles (4 spindles for L model, and 6 spindles for R and H models), viscometer stand and spindle protector, carrying case, USB cable, power supply cable and Datalogger Software.