Digital Rotational Viscometer ALPHA Series, Fungilab
Supplier: Fungilab
V100001 V101001 V100002 V101002
89236-996EA 5238.89 USD
89236-996 89236-998 89237-178 89237-176
Digital Rotational Viscometer ALPHA Series, Fungilab
Viscometers Rotational Viscometers
Alpha Series is a basic digital rotational viscometer for easy viscosity readings
Main features include: 4LCD display for easy menu access and results reading, relative and absolute viscosity reading automatically, unit convert SI to CGS; Autotest and Autorange functions; 10 language options. 6 touch keypad. LCD screen. 4 lines.
Low budget and easy to use.
Ordering information: Standard delivery includes:head boss, standard spindles (6 spindles for R&H models / 4 spindles for L models), viscometer stand, spindle protector and user's manual. Carrying case not included. No software available. No temperature function.