Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BruClean™ Disinfectant Tablet, ITW Texwipe

Supplier: ITW-TEXWIPE COMPANY
TX6466
Bru-Clean is a premeasured tablet that, when added to one gallon of water, creates a disinfectant cleaner for hard surfaces that is effective against a broad array of pathogens, including MRSA

The active ingredient, sodium dichloroisocyanurate (NaDCC), features 1) Stability: more stable than bleach (sodium hypochlorite), 2) Neutral pH: advanced formula is almost neutral in pH, 3) Protects Stainless Steel: much less corrosive than bleach on stainless steel and other metals, and 4) biodegradable. Strength: Concentration of 937 ppm over time available chlorine.

1000 ppm available chlorine for 24 hours. Pre-scored tablets. Product is EPA-registered (71847-2-106). Not Hazardous.

Save on shipping and storage costs by purchasing lightweight, dry tablets instead of heavy, liquid bleach.

Certifications: EPA Registered.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Bru-Clean Disinfectant Tablets Bulk packed tablets TX6466 TWTX6466 Case of 540 Retrieving Restricted
Pack of 270 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Stability Made at point-of-use
pH 5-6 Alkaline
Shelf Life Shelf life of 2 years
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty