Xylenol orange 0.1% (w/v) in alcohol USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
R9387000-120A
R9387000-500A
RC9387-16EA
238.88
USD
RC9387-16
RC9387-4
Xylenol orange 0.1% (w/v) in alcohol USP Test Solution (TS)
Xylenol orange
Formula:
C₃₁H₂₈N₂Na₄O₁₃S MW: 760.59 g/mol |
MDL Number:
MFCD00044293 CAS Number: 3618-43-7 UN: 1987 ADR: 3,II |
Specification Test Results
Appearance | Clear, red, alcohol odor |
Suitability for Use | Purple (Excess Metal) - Yellow (Excess EDTA) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...