Tetraethylammonium bromide 40% (w/w) in glacial acetic acid
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Tetrylammonium bromide
R8385000-1C
RC8385-32EA
788.7
USD
RC8385-32
Tetraethylammonium bromide 40% (w/w) in glacial acetic acid
Tetraethylammonium bromide
Formula:
C₈H₂₀BrN MW: 210.16 g/mol Boiling Pt: ~ 118 °C (Acetic Acid) Density: 1.0500 g/cm³ Flash Pt: 39 °C (AcOH) Storage Temperature: Ambient |
MDL Number:
MFCD00011825 CAS Number: 71-91-0 UN: 2790 ADR: 8,II |
Specification Test Results
Appearance | Clear, tan-gold, Strong Acetic odor |
Assay at 20 °C (traceable to NIST SRM 999) | 40 ± 1 % (w/w) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...