Tin(II) chloride 10% (w/v) in 0.5 N sulfuric acid, suspension
Supplier: Ricca Chemical
Synonyms:
Stannous chloride
R7996500-1C
RC7996.5-32EA
274.48
USD
RC7996.5-32
Tin(II) chloride 10% (w/v) in 0.5 N sulfuric acid, suspension
Tin (II) chloride
Formula:
SnCl₂ MW: 189.62 g/mol |
MDL Number:
MFCD00011241 CAS Number: 7772-99-8 UN: 3260 ADR: 8,III |
Specification Test Results
Appearance | Cloudy, suspension, odorless |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...