SPADNS reference solution for fluoride determination
Supplier: Ricca Chemical
|
Danger
|
R7990000-500C
RC799016EA
63.97
USD
RC799016
SPADNS reference solution for fluoride determination
SPADNS reference solution
Boiling Pt:
~ 100 °C Density: 1.0130 g/cm³ Storage Temperature: Ambient |
UN:
3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, red, slightly pungent |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...