Sodium tungstate 10% (w/v) in aqueous solution stabilized

Supplier: Ricca Chemical
Warning

R7980000-500A
RC798016EA 73.59 USD
RC798016
Sodium tungstate 10% (w/v) in aqueous solution stabilized
Sodium tungstate solutions
Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0900 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Specific Gravity 1.09 ± 0.02 sp gr

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR