Disodium sulfide 3% (w/v) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Sodium sulphideSodium sulfide

R7570000-1C R7570000-500C
RC7570-16EA 112.74 USD
RC7570-16 RC7570-32
Disodium sulfide 3% (w/v) in aqueous solution
Disodium sulphide
Formula: Na₂S
MW: 78.05 g/mol
MDL Number: MFCD00003498
CAS Number: 1313-82-2
UN: 1385
ADR: 4.2,II

Order Now

Specification Test Results

Appearance Clear, colorless, Sulfide odor
Refractive Index Confirmed Concentration

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR