Sodium metaarsenite 0.100 M (M/10)
Supplier: Ricca Chemical
R7142000-1C
R7142000-500C
RC7142-32EA
108.77
USD
RC7142-32
RC7142-16
Sodium metaarsenite 0.100 M (M/10)
Sodium metaarsenite
Formula:
NaAsO₂ MW: 129.91 g/mol |
MDL Number:
MFCD00003472 CAS Number: 7784-46-5 UN: 2027 ADR: 6.1,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 83) | 0.1000 ± 0.0001 M at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...