Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, RICCA Chemical Company

Supplier: Ricca Chemical
R7109750-500A
RC7109.75-16EA 102.05 USD
RC7109.75-16
Simulated Intestinal Fluid (without Pancreatin), USP XXII Formulation, RICCA Chemical Company
Standards Biological Standards
Appearance: Colorless liquid
pH at 25°C (Method: SQCP027, SQCP033) 7.40-7.60
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR