Diammonium hydrogen orthophosphate 13% (w/v) in aqueous solution USP Test Solution (TS), USP Reference Solution (RS)
Supplier: Ricca Chemical
|
Warning
|
Synonyms:
Ammonium Phosphate
, di-Ammonium hydrogen phosphate
, Diammonium hydrogen orthophosphate
, Ammonium hydrogen phosphate
, Ammonium monohydrogen phosphate
, Ammonium phosphate dibasic
R0686000-500A
RC686-16EA
117.4
USD
RC686-16
Diammonium hydrogen orthophosphate 13% (w/v) in aqueous solution USP Test Solution (TS), USP Reference Solution (RS)
di-Ammonium hydrogen orthophosphate
Formula:
(NH₄)₂HPO₄ MW: 132.06 g/mol Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0600 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Refractive Index | Confirmed concentration |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...