di-Ammonium oxalate 3.5% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Warning

R0679000-500A R0679000-1A
RC679-16EA 81.24 USD
RC679-16 RC679-32
di-Ammonium oxalate 3.5% (w/v) in aqueous solution USP Test Solution (TS)
di-Ammonium oxalate solutions
Formula: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40) 3.50 ± 0.03 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR