Dipotassium oxalate 2% (w/v) in aqueous solution
Supplier: Ricca Chemical
R6320000-500A
R6320000-1A
RC6320-32EA
85.57
USD
RC6320-32
RC632016
Dipotassium oxalate 2% (w/v) in aqueous solution
di-Potassium oxalate
Formula:
C₂K₂O₄ MW: 166.22 g/mol |
CAS Number:
583-52-8 UN: 3282 ADR: 6.1,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Refractive Index | 2.0 ± 0.3 % (w/v) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...