Ammonium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
Synonyms:
Acetic acid ammonium salt
R0618000-1A
R0618000-500A
RC618-16EA
60.2
USD
RC618-16
RC618-32
Ammonium acetate 10% (w/v) in aqueous solution USP Test Solution (TS)
Ammonium acetate
Formula:
H₃CCOONH₄ MW: 77.08 g/mol Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0200 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Specific Gravity | 10.0 ± 1.2 % (w/v) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...