Ammonia, Dilute R1 102 g/L

Supplier: Ricca Chemical
Danger

R0610200-500A
RC610.2-16EA 125.53 USD
RC610.2-16
Ammonia, Dilute R1 102 g/L
Ammonia, Dilute R1
Formula: H3N
MW: 17,03 g/mol
Boiling Pt: ~ 36 °C
Melting Pt: ~ 0 °C
Density: 0.9555 g/cm³
Storage Temperature: Ambient
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, ammonia odor
Assay at 20 °C (traceable to NIST SRM 723) 102 ± 2 g/L NH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR