Phenylarsine oxide 0.0375 N stabilized titrant

Supplier: Ricca Chemical

Synonyms: OxophenylarsineArzenePAO

R5760000-1C
RC576032EA 598.96 USD
RC576032
Phenylarsine oxide 0.0375 N stabilized titrant
Phenylarsine oxide
Formula: C₆H₅AsO
MW: 168.02 g/mol
MDL Number: MFCD00001990
CAS Number: 637-03-6
UN: 3465
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear to hazy, gold to light yellow, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.03750 ± 0.00002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR