Phenanthroline ferrous sulfate (orthophenanthroline TS) 0.025 M
Supplier: Ricca Chemical
R5464800-120C
R5464800-500C
RC5464.8-4EA
99.61
USD
RC5464.8-4
RC5464.8-16
Phenanthroline ferrous sulfate (orthophenanthroline TS) 0.025 M
Phenanthroline ferrous sulfate (orthophenanthroline TS)
Mixture 1,10-Phenanthroline Monohydrate, Ferrous Sulfate Heptahydrate in aqueous solution.
Specification Test Results
Appearance | brownish orange to blood red, odorless |
Suitability for Use | Green to Bluish Green (Reduced) - Redish (Oxidized) |