Ammonium iron (II) sulfate 0.0500 N (N/20)

Supplier: Ricca Chemical
Danger

R3143000-500C R3143000-1C
RC3143-32EA 122.53 USD
RC3143-32 RC3143-16
Ammonium iron (II) sulfate 0.0500 N (N/20)
Haematoxylin Harris
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9

Order Now

Specification Test Results

Appearance Clear, light green, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.05000 ± 0.00005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR