Ammonium iron (III) sulfate 0.125 N (N/8)

Supplier: Ricca Chemical
Warning

R3062000-20F
RC3062-5EA 938 USD
RC3062-5
Ammonium iron (III) sulfate 0.125 N (N/8)
Ammonium iron(III) sulphate solutions
Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0277 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7

Order Now

Specification Test Results

Appearance Clear, yellow, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.1250 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR