1,5-Diphenylcarbazone solution 0.1% (w/v) alcoholic solution indicator

Supplier: Ricca Chemical
Danger

Synonyms: sym-DiphenylcarbazonePhenylazoformic acid 2-phenylhydrazidPhenylazoameisensäure-2-phenylhydrazidPhenylazoformic acid 2-phenylhydrazide

R2600000-500C
RC260016EA 140.3 USD
RC260016
1,5-Diphenylcarbazone solution 0.1% (w/v) alcoholic solution indicator
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003024
CAS Number: 538-62-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear. Orange, alcohol odor
Suitability for Use Colorless (excess Chloride) - Purple (excess Mercury)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR