Ethyl-2-aminobenzo[b]thiophene-3-carboxylate for synthesis, Sigma-Aldrich®
Synonyms:
Ethyl 2-aminobenzo[b]thiophene-3-carboxylate
M-Clarity™ quality level = MQ200
8.14873.0001
EM8.14873.0001EA
228.16
USD
EM8.14873.0001
Ethyl-2-aminobenzo[b]thiophene-3-carboxylate for synthesis, Sigma-Aldrich®
Ethyl 2-aminobenzo[b]thiophene-3-carboxylate
Formula:
C₁₁H₁₁NO₂S MW: 221.2755 g/mol |
MDL Number:
MFCD00102506 CAS Number: 7311-95-7 UN: 2811 ADR: 6.1,II |
Specification Test Results
Assay (HPLC, area%) | 96% min |
Identity | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...