Nuclear Fast Red solution stable stain for histology

Synonyms: Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonateSodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulfonateCI 60760Helio Fast Rubin BBL

VWRV1B1369-500ML
89230-006EA 159.86 USD
89230-006
Nuclear Fast Red solution stable stain for histology
Nuclear Fast Red

  • Ready-to-use solution
  • No precipitation upon storage
  • No filtering required


VWR’s Liquid-Stable Nuclear Fast Red is a room temperature, stable solution formulated specifically to prevent precipitation during storage, which is a problem commonly encountered in homemade solutions and competitors’ products. Liquid-Stable Nuclear Fast Red is used in immunohistochemistry as a nuclear counterstain on slides treated with routine histological chromogens or stains, such as DAB or Prussion blue. Light red or pink staining of nuclei requires only a few minutes of incubation in Liquid-Stable Nuclear Fast Red following the primary staining step. Slides are then washed in water, dehydrated and cleared before mounting with an organic mounting medium.

Formula: C₁₄H₈NNaO₇S
MW: 357.28 g/mol
MDL Number: MFCD00078493
CAS Number: 6409-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR