Would you like to log in again?
VWR Life Science AMRESCO BCS Assay Kit
Supplier: VWR
  • Reduced variability from protein to protein sample
  • High tolerance for salts, detergents and buffers
  • Reproducible and Consistent Results
The BCS Assay is a kit that allows for spectrophotometric determination of protein concentration. The BCS Assay utilizes a biuret-like reaction where Copper (II) binds protein. The remaining free Copper (II) then is reduced to Copper (I) which associates with a dye that changes colors upon binding of copper (I). Protein concentration is then determined by measuring the absorbance of the dye-Copper (I) complex. The signal is inversely proportional to the concentration of peptide bonds as opposed to specific amino acid side chains. Therefore, protein to protein variability is less of a concern. Another advantage of the BCS Assay is that the BCS assay has a high tolerance for salts, detergents, and buffers that may interfere with conventional protein quantitation kits.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Size Application Supplier No. VWR Catalog Number Unit Price Quantity
BCS Assay Kit Kit Sufficient material for 100 assays. N962-100RXN 71005-968 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty